SYRINGE FILTER MCE 0.22UM 13MM S CS75
Catalog # 77973-128
Supplier: VistaLab
New Product
SYRINGE FILTER MCE 0.22UM 13MM S CS75
Catalog # 77973-128
Supplier: VistaLab
Supplier Number: 229750
New Product
Frequently Bought Together
VWR® TraceClean® Wide Mouth Jars, HDPE
Supplier: VWR International
VWR® Test Tube Racks
Supplier: VWR International
VWR® Cryo Racks for 1.0, 1.8 and 4.5 ml Cryogenic Tubes, PC
Supplier: VWR International
VWR® Single Scale Graduated Cylinders, PP
Supplier: VWR International
SP Bel-Art Tall Form Graduated Pitchers, Polypropylene, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
SP Bel-Art Poxygrid® Labware Drying Rack, Epoxy-Coated Steel Wire, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
BowTie™ Biohazard Bag and Bin
Supplier: MTC BIO, INC.
Nalgene® Unwire™ Half-Racks, ResMer® Resin, Thermo Scientific
Supplier: Thermo Fisher Scientific
VWR® Freezer Storage Boxes, 16-Place and 36-Place
Supplier: VWR International
VWR® 80-Well Microtube Racks
Supplier: VWR International
VWR® TPX Measuring Cylinders, Class B
Supplier: VWR International
VWR® TraceClean® Cylinder Bottles, HDPE
Supplier: VWR International
SP Bel-Art No-Wire™ Multi-Tube Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
VITLAB® HDPE Buckets, BrandTech
Supplier: Brandtech
Plastic Packers, Wide Mouth, HDPE, Natural, C&G Containers
Supplier: C&G Containers
VWR® TraceClean® Cylinders, Narrow Mouth, HDPE, Natural
Supplier: VWR International
VWR® TraceClean® Packer Containers, HDPE, Wide Mouth, Natural
Supplier: VWR International