CELL STRAINER 100UM BULK STERILE CS50
Catalog # 77972-878
Supplier: VistaLab
New Product
CELL STRAINER 100UM BULK STERILE CS50
Catalog # 77972-878
Supplier: VistaLab
Supplier Number: 229486
New Product
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® High Performance Microcentrifuge Tubes, PP
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
Eppendorf® Safe-Lock Tubes, 1.5 ml - Microtubes
Supplier: Eppendorf
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: Eppendorf
Screw Cap Tubes, 5.0 ml, MTC Bio
Supplier: GeneMate
Micrewtube™ Microcentrifuge Tubes, Simport Scientific
Supplier: Simport Scientific
VWR® Microcentrifuge Tubes with Socket Screw-Cap
Supplier: VWR International
VWR® Disposable Aspirating Pipettes, Polystyrene, Sterile
Supplier: VWR International
VWR® Disposable Microcentrifuge Tubes
Supplier: VWR International
LoBind Protein or Genomic Microcentrifuge Tubes, Eppendorf
Supplier: Eppendorf
Nalgene® Cryo 1°C 'Mr. Frosty' Freezing Container, Thermo Scientific
Supplier: Thermo Fisher Scientific
VWR® Micro Centrifuge Tube with Flat Screw-Cap
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International