RACK 80 WELL MCT TUBE CS20
Catalog # 77972-202
Supplier: POLYCARBIN, INC.
New Product
RACK 80 WELL MCT TUBE CS20
Catalog # 77972-202
Supplier: POLYCARBIN, INC.
Supplier Number: 2PR80
New Product
Frequently Bought Together
Tube Rack, One-Well Wire, for 50 ml Tubes
Supplier: Heathrow Scientific
Cooling Containers
Supplier: Heathrow Scientific
VWR® TraceClean® Wide Mouth Jars, HDPE
Supplier: VWR International
VWR® Microtube Storage Boxes
Supplier: VWR International
VWR® Test Tube Racks
Supplier: VWR International
VWR® Cryogenic Fiberboard Freezer Box
Supplier: VWR International
VWR® Cuvette Racks, PP
Supplier: VWR International
Mega Tube Racks for 18 - 20 mm Tubes
Supplier: Heathrow Scientific
Y-Shaped Connectors
Supplier: Dynalon
PolarSafe™ Polypropylene Storage Boxes, 15 and 50 ml, Argos Technologies
Supplier: Antyila Scientific
BowTie™ Biohazard Bag and Bin
Supplier: MTC BIO, INC.
Fold and Snap Tube Racks, 21 mm, 40-Place
Supplier: VWR International
REUZ™ Stainless Steel Beakers with Handle
Supplier: United Scientific Supplies
Jar with Cap, HDPE
Supplier: Dynalon
SP Bel-Art Poxygrid® Labware Drying Rack, Epoxy-Coated Steel Wire, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
VWR® Flexible Safety Caps
Supplier: VWR International
VWR® Pop-Up™ Tube Racks
Supplier: VWR International
VWR® TraceClean® Wide Mouth Packers, HDPE
Supplier: VWR International