Basic Latent Print Kits
Standard fingerprint kit, orange case
Catalog # 470358-948
New Product
Supplier: THE LYNN PEAVEY COMPANY
Supplier #: 40023
Jump to:
Product Details & Documents
Great for classroom use.
Full set of materials in a single pack
- Standard black or high visibility orange storage box
Use for lifting latent fingerprints from standard non-porous surfaces like glass, and smooth painted surfaces. Each kit includes a case with a foam liner to keep backers, tape, brush, and powder separated.
Specifications
- Description:Standard fingerprint kit, orange case