RECOMBINANT HUMAN MK-3 PROT AB60308-5UG
ABCA_AB60308-5UG
: Abcam
New Product
Frequently Bought Together
BUFFER JAW SSC 2X 1L PACKS
: G-Biosciences
HTRF STREPTAVIDIN TB-CONJUGATE 1000ASSAY
: Revvity Health Sciences
72192 PROSTAGLANDIN E2 1MG
: STEMCELL Technologies
Chicken Embryo Extract, MP Biomedical
: MP Biomedicals
Fixation, (buffer), Enzo Life Sciences
: Enzo Life Sciences
Canine Plasma, Equitech- Bio, Inc.
: Equitech-Bio
Bovine Plasma with Citrate, Equitech- Bio, Inc.
: Equitech-Bio
Nano-Glo® Luciferase Assay System, Promega, Promega
: Promega Corporation
74042 PHORBOL12 MYRISTATE 13 ACETATE 1MG
: STEMCELL Technologies
KIT ELISA ALT COMPLEMNT PATHWY 100UL/WEL
: Hycult Biotech
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
: Invitrogen
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
: Meso Scale Diagnostics
Rat Plasma, Equitech- Bio
: Equitech-Bio
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
: Genetex
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
: Invitrogen
ASSAY IMMUNO CEFRAS GLOBAL CD8 HU 2VIAL
: MABTECH INC MS
Fixation/Permeabilization, (buffer), Enzo Life Sciences
: Enzo Life Sciences
73254 METFORMIN HYDROCHLORIDE 5G
: STEMCELL Technologies