RECOMBINANT HUMAN PRAK/MK AB217826-20UG
Catalog # ABCA_AB217826-20UG
Supplier: Abcam
New Product
RECOMBINANT HUMAN PRAK/MK AB217826-20UG
Catalog # ABCA_AB217826-20UG
Supplier: Abcam
Supplier Number: AB217826-20UG
New Product
Frequently Bought Together
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
Bovine Plasma with Citrate, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
100 1042 CHIR99021
Supplier: STEMCELL Technologies
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
Supplier: Invitrogen
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Human Recombinant Shh (C24II)
Supplier: STEMCELL Technologies
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
Supplier: Meso Scale Diagnostics
Rat Plasma, Equitech- Bio
Supplier: Equitech-Bio
100 0544 ERASTIN
Supplier: STEMCELL Technologies
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
Goat Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
Supplier: Invitrogen
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
LIGAND 646 HALOTAG 5UG LMAGING LIVE CELL
Supplier: Promega Corporation
KIT HUMAN GIPR REPORTER 3X32ASSAYS 96WL
Supplier: INDIGO BIOSCIENCES INC MS
HTRF STREPTAVIDIN TB-CONJUGATE 1000ASSAY
Supplier: Revvity Health Sciences