Order Entry
United States
Orders LinkContactUsLinkComponent
Hygromycin B ≥90%

Hygromycin B ≥90%

500 mg
Catalog # ABCA_AB286934-500M
New Product
Supplier:   Abcam
Supplier #:  AB286934-500MG
Hygromycin B ≥90%
Catalog # ABCA_AB286934-500M
Supplier:  Abcam
New Product
CAS Number:  31282-04-9
Hygromycin B ≥90%

500 mg

CAS Number:  31282-04-9
Price
$211.00
Log in for availability
Each (500mg)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

An aminoglycoside antibiotic produced by Streptomyces hygroscopicus that is routinely used as a selection agent for cells that contain hph or hyg Hygromycin resistant gene. Hygromycin B is thought to inhibit protein synthesis by strengthening the interaction of peptidyl-tRNA in the ribosomal acceptor ('A') site thereby preventing translocation of by EF-2 (elongation Factor-2). Hygromycin B is effective on most bacteria, fungi and higher eukaryotes.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol
  • CAS Number:31282-04-9
  • Molecular Weight:527.5 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Hygromycin B

Specifications

  • Size:
    500 mg

Specifications