Order Entry
United States
Orders LinkContactUsLinkComponent
 

Polysorbate 80 solution 0,1

100 ml
Catalog # ABCA_AB291111-100M
New Product
Supplier:   Abcam
 :  AB291111-100ML
Polysorbate 80 solution 0,1
Catalog # ABCA_AB291111-100M
Supplier:  Abcam
New Product
CAS Number:  9005-65-6
Polysorbate 80 solution 0,1

100 ml

CAS Number:  9005-65-6
Price
$180.00
Log in for availability
Each (100ml)
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Tween 80 Detergent, 10% Solution, ab291111 (previously known as Tween® 80, MegaPure™ Detergent, 10% Solution, 2126) is a non-ionic detergent useful for the extraction of membrane-bound proteins as well as isolation of nuclei from mammalian cell lines.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate
  • CAS Number:9005-65-6
Specifications

Specification Test Results

  • Constituents::
    10% Tween 80

Specifications

  • Size:
    100 ml

Specifications