ANTIBODY POLR2A A322799 100UL
Catalog # 77971-306
Supplier: ANTIBODIES.COM LLC
New Product
ANTIBODY POLR2A A322799 100UL
Catalog # 77971-306
Supplier: ANTIBODIES.COM LLC
Supplier Number: A322799-100
New Product
Frequently Bought Together
Bovine Plasma with Citrate, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
Supplier: Meso Scale Diagnostics
100 0544 ERASTIN
Supplier: STEMCELL Technologies
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
Supplier: Invitrogen
Goat Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
ASSAY IMMUNO CEFRAS GLOBAL CD8 HU 2VIAL
Supplier: MABTECH INC MS
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
BLOCKER AQB013 AQUAPORIN 5MG
Supplier: FOCUS BIOMOLECULES MS
KIT HUMAN GIPR REPORTER 3X32ASSAYS 96WL
Supplier: INDIGO BIOSCIENCES INC MS
MOUSE SERUM US ORIGIN FOR COGLTN STUDIES
Supplier: Innovative Research Inc
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
ASSAY TAQMAN/ GENE EXPRESSION FAM 75RXN
Supplier: Invitrogen
73254 METFORMIN HYDROCHLORIDE 5G
Supplier: STEMCELL Technologies
KIT HUMAN GLP-1R REPORTER 3X32ASSAYS
Supplier: INDIGO BIOSCIENCES INC MS
KIT ELISA ALT COMPLEMNT PATHWY 100UL/WEL
Supplier: Hycult Biotech
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
Supplier: Invitrogen