ANTIBODY ANTI-MAGEA4 OTI1F9 322366 100UG
77970-930
New Product
Frequently Bought Together
DEFIBRINATED BLOOD SHEEP 100ML
: LAMPIRE BIOLOGICAL LAB MS
Chicken Embryo Extract, MP Biomedical
: MP Biomedicals
HTRF STREPTAVIDIN TB-CONJUGATE 1000ASSAY
: Revvity Health Sciences
BUFFER JAW SSC 2X 1L PACKS
: G-Biosciences
LIGAND 646 HALOTAG 5UG LMAGING LIVE CELL
: Promega Corporation
Nano-Glo® Luciferase Assay System, Promega, Promega
: Promega Corporation
Bovine Plasma with Citrate, Equitech- Bio, Inc.
: Equitech-Bio
Canine Plasma, Equitech- Bio, Inc.
: Equitech-Bio
Fixation, (buffer), Enzo Life Sciences
: Enzo Life Sciences
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
: Invitrogen
Human Recombinant Shh (C24II)
: STEMCELL Technologies
100 0544 ERASTIN
: STEMCELL Technologies
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
: Meso Scale Diagnostics
72262 ALL TRANS RETINOIC ACID 50MG
: STEMCELL Technologies
Fixation/Permeabilization, (buffer), Enzo Life Sciences
: Enzo Life Sciences
ASSAY TAQMAN/ GENE EXPRESSION FAM 75RXN
: Invitrogen
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
: Genetex
Goat Plasma, Equitech- Bio, Inc.
: Equitech-Bio
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
: Invitrogen
AGONIST RESINIFERATOXIN TRPV1 100UG
: FOCUS BIOMOLECULES MS