ANTIBODY HSP90 BETA A322210 100UL
Catalog # 77970-788
Supplier: ANTIBODIES.COM LLC
New Product
ANTIBODY HSP90 BETA A322210 100UL
Catalog # 77970-788
Supplier: ANTIBODIES.COM LLC
Supplier Number: A322210-100
New Product
Frequently Bought Together
VWR® Stripping Buffers
Supplier: VWR
Mix & Go!™ E. coli Transformation Kit and Buffer Set
Supplier: Zymo Research
HTRF STREPTAVIDIN TB-CONJUGATE 1000ASSAY
Supplier: Revvity Health Sciences
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
TAE 50X Buffer, MP Biomedicals
Supplier: MP Biomedicals
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Radiance Q Chemiluminescent Substrate, Azure Biosystems
Supplier: Azure Biosystems
Tris HCl pH 8.5, 1.0 M
Supplier: Rockland Immunochemical
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
Supplier: Invitrogen
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
73254 METFORMIN HYDROCHLORIDE 5G
Supplier: STEMCELL Technologies
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
Supplier: Meso Scale Diagnostics
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
Supplier: Invitrogen