ANTIBODY ANTI-OBFC2A OTI6B10 100UL
Catalog # 77971-095
Supplier: ANTIBODIES.COM LLC
New Product
ANTIBODY ANTI-OBFC2A OTI6B10 100UL
Catalog # 77971-095
Supplier: ANTIBODIES.COM LLC
Supplier Number: A322558-100
New Product
Frequently Bought Together
72192 PROSTAGLANDIN E2 1MG
Supplier: STEMCELL Technologies
Bovine Plasma with Citrate, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
KIT BD-TAU ADVANTAGE PLUS
Supplier: QUANTERIX CORPORATION MS
PI MARKER ICE280/ICE3 6.61 200UL
Supplier: 0000014126
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT HUMAN GLP-1R REPORTER 3X32ASSAYS
Supplier: INDIGO BIOSCIENCES INC MS
ASSAY TAQMAN/ GENE EXPRESSION FAM 75RXN
Supplier: Invitrogen
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
Supplier: Invitrogen
Human Recombinant Shh (C24II)
Supplier: STEMCELL Technologies
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
Supplier: Meso Scale Diagnostics
Rat Plasma, Equitech- Bio
Supplier: Equitech-Bio
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
ASSAY IMMUNO CEFRAS GLOBAL CD8 HU 2VIAL
Supplier: MABTECH INC MS
Goat Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
Supplier: Invitrogen
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
100 0251 ELTROMBOPAG 5MG
Supplier: STEMCELL Technologies
KIT HUMAN GIPR REPORTER 3X32ASSAYS 96WL
Supplier: INDIGO BIOSCIENCES INC MS