ANTIBODY ANTI-MAN1 OTI6C10 A322370 100UG
Catalog # 77970-934
Supplier: ANTIBODIES.COM LLC
New Product
ANTIBODY ANTI-MAN1 OTI6C10 A322370 100UG
Catalog # 77970-934
Supplier: ANTIBODIES.COM LLC
Supplier Number: A322370-100
New Product
Frequently Bought Together
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
72262 ALL TRANS RETINOIC ACID 50MG
Supplier: STEMCELL Technologies
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
Supplier: Invitrogen
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
MOUSE SERUM US ORIGIN FOR COGLTN STUDIES
Supplier: Innovative Research Inc
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Bovine Plasma with Citrate, Equitech- Bio, Inc.
Supplier: Equitech-Bio
KIT BD-TAU ADVANTAGE PLUS
Supplier: QUANTERIX CORPORATION MS
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
74042 PHORBOL12 MYRISTATE 13 ACETATE 1MG
Supplier: STEMCELL Technologies
100 1042 CHIR99021
Supplier: STEMCELL Technologies
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
ASSAY IMMUNO CEFRAS GLOBAL CD8 HU 2VIAL
Supplier: MABTECH INC MS
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
Supplier: Invitrogen
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
Supplier: Meso Scale Diagnostics