ANTIBODY KLF5 OTI2G4 A322300 100UG
Catalog # 77970-871
Supplier: ANTIBODIES.COM LLC
New Product
ANTIBODY KLF5 OTI2G4 A322300 100UG
Catalog # 77970-871
Supplier: ANTIBODIES.COM LLC
Supplier Number: A322300-100
New Product
Frequently Bought Together
DEFIBRINATED BLOOD SHEEP 100ML
Supplier: LAMPIRE BIOLOGICAL LAB MS
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
MARKER MAURICE CIEF PI-8.40 LYOPH 210UL
Supplier: 0000014126
73884 DIBUTYRYL CAMP SODIUM SALT 100MG
Supplier: STEMCELL Technologies
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
MOUSE SERUM US ORIGIN FOR COGLTN STUDIES
Supplier: Innovative Research Inc
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
LIGAND 646 HALOTAG 5UG LMAGING LIVE CELL
Supplier: Promega Corporation
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
Supplier: Invitrogen
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
Supplier: Meso Scale Diagnostics
Rat Plasma, Equitech- Bio
Supplier: Equitech-Bio
72262 ALL TRANS RETINOIC ACID 50MG
Supplier: STEMCELL Technologies
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
BUFFER STABILIZR 7.5% DTPA-PURI BSA 50ML
Supplier: Revvity Health Sciences
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
ASSAY IMMUNO CEFRAS GLOBAL CD8 HU 2VIAL
Supplier: MABTECH INC MS
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
Supplier: Invitrogen