ANTIBODY KLF2 OTI5F1 A322298 100UG
Catalog # 77970-869
Supplier: ANTIBODIES.COM LLC
New Product
ANTIBODY KLF2 OTI5F1 A322298 100UG
Catalog # 77970-869
Supplier: ANTIBODIES.COM LLC
Supplier Number: A322298-100
New Product
Frequently Bought Together
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
72192 PROSTAGLANDIN E2 1MG
Supplier: STEMCELL Technologies
HTRF STREPTAVIDIN TB-CONJUGATE 1000ASSAY
Supplier: Revvity Health Sciences
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
LIGAND 646 HALOTAG 5UG LMAGING LIVE CELL
Supplier: Promega Corporation
MOUSE SERUM US ORIGIN FOR COGLTN STUDIES
Supplier: Innovative Research Inc
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Bovine Plasma with Citrate, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
100 1042 CHIR99021
Supplier: STEMCELL Technologies
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
Supplier: Meso Scale Diagnostics
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
ASSAY IMMUNO CEFRAS GLOBAL CD8 HU 2VIAL
Supplier: MABTECH INC MS
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
Supplier: Invitrogen
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
73254 METFORMIN HYDROCHLORIDE 5G
Supplier: STEMCELL Technologies
72262 ALL TRANS RETINOIC ACID 50MG
Supplier: STEMCELL Technologies
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
Supplier: Invitrogen