ANTIBODY ANTI-KI67 UMAB107 A322296 100UL
Catalog # 77970-868
Supplier: ANTIBODIES.COM LLC
New Product
ANTIBODY ANTI-KI67 UMAB107 A322296 100UL
Catalog # 77970-868
Supplier: ANTIBODIES.COM LLC
Supplier Number: A322296-100
New Product
Frequently Bought Together
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT HUMAN GLP-1R REPORTER 3X32ASSAYS
Supplier: INDIGO BIOSCIENCES INC MS
73254 METFORMIN HYDROCHLORIDE 5G
Supplier: STEMCELL Technologies
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
100 0251 ELTROMBOPAG 5MG
Supplier: STEMCELL Technologies
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
SOLUTION ATTUNE WASH 250ML
Supplier: Invitrogen
KIT ELISA ALT COMPLEMNT PATHWY 100UL/WEL
Supplier: Hycult Biotech
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
Supplier: Invitrogen
100 0544 ERASTIN
Supplier: STEMCELL Technologies
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
Supplier: Meso Scale Diagnostics
Rat Plasma, Equitech- Bio
Supplier: Equitech-Bio
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
Goat Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
Supplier: Invitrogen
AGONIST RESINIFERATOXIN TRPV1 100UG
Supplier: FOCUS BIOMOLECULES MS
ASSAY IMMUNO CEFRAS GLOBAL CD8 HU 2VIAL
Supplier: MABTECH INC MS