ELISA KIT MOUSE RANKL A3634 96 TEST
Catalog # 77967-424
Supplier: ANTIBODIES.COM LLC
New Product
ELISA KIT MOUSE RANKL A3634 96 TEST
Catalog # 77967-424
Supplier: ANTIBODIES.COM LLC
Supplier Number: A3634-96
New Product
Frequently Bought Together
VWR® Stripping Buffers
Supplier: VWR
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
Pharmalytes® IEF Carrier Ampholytes, Cytiva
Supplier: Cytiva
Amersham ECL Western Blotting Select Detection Reagent, Cytiva
Supplier: Cytiva
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
LIGAND 646 HALOTAG 5UG LMAGING LIVE CELL
Supplier: Promega Corporation
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Radiance Q Chemiluminescent Substrate, Azure Biosystems
Supplier: Azure Biosystems
PI MARKER 9.50 200UL
Supplier: 0000014126
Tris HCl pH 8.5, 1.0 M
Supplier: Rockland Immunochemical
DNA Wash Buffer, Concentrate, Zymo Research
Supplier: Zymo Research
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Chemi Blot Blocking Buffer, Azure
Supplier: Azure Biosystems
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex