ELISA KIT RAT PACAP-38 A3667 48 TEST
Catalog # 77968-067
Supplier: ANTIBODIES.COM LLC
New Product
ELISA KIT RAT PACAP-38 A3667 48 TEST
Catalog # 77968-067
Supplier: ANTIBODIES.COM LLC
Supplier Number: A3667-48
New Product
Frequently Bought Together
VWR® Stripping Buffers
Supplier: VWR
Sodium chloride 5 M, sterile-filtered
Supplier: Teknova
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
Mix & Go!™ E. coli Transformation Kit and Buffer Set
Supplier: Zymo Research
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Tris HCl pH 8.5, 1.0 M
Supplier: Rockland Immunochemical
DNA Wash Buffer, Concentrate, Zymo Research
Supplier: Zymo Research
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
LIGAND 646 HALOTAG 5UG LMAGING LIVE CELL
Supplier: Promega Corporation
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
FACS and IF Staining Buffer with BSA
Supplier: Rockland Immunochemical
Proteomelab™ SDS-MW Analysis Kit, SCIEX
Supplier: 0000013719
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex