EDTA SOLUTION (0.05%) IN PBS 100ML
Catalog # 77968-460
Supplier: Sartorius
New Product
EDTA SOLUTION (0.05%) IN PBS 100ML
Catalog # 77968-460
Supplier: Sartorius
Supplier Number: 03-015-1B
New Product
Frequently Bought Together
L(+)-Cysteine hydrochloride monohydrate, high purity
Supplier: VWR
Bovine Plasma with Citrate, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Potassium chloride
Supplier: G-Biosciences
MOUSE SERUM US ORIGIN FOR COGLTN STUDIES
Supplier: Innovative Research Inc
Sodium acetate
Supplier: G-Biosciences
MARKER 6.14 MAURICE CIEF PI 210UL
Supplier: 0000014126
DEFIBRINATED BLOOD SHEEP 100ML
Supplier: LAMPIRE BIOLOGICAL LAB MS
Research Buffers and Reagents, RNase and DNase free, G-Biosciences
Supplier: G-Biosciences
Sodium chloride 0.9% in water normal saline solution
Supplier: G-Biosciences
Magnesium chloride hexahydrate for biochemistry
Supplier: G-Biosciences
Wax Paper Sheets, Cytiva
Supplier: Cytiva
Goat Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Sodium chloride
Supplier: G-Biosciences
Magnesium chloride hexahydrate cell culture reagent
Supplier: Thermo Scientific Chemicals
Rat Plasma, Equitech- Bio
Supplier: Equitech-Bio