ELISA KIT BOVINE MCP2 A4339 48 TEST
Catalog # 77967-147
Supplier: ANTIBODIES.COM LLC
New Product
ELISA KIT BOVINE MCP2 A4339 48 TEST
Catalog # 77967-147
Supplier: ANTIBODIES.COM LLC
Supplier Number: A4339-48
New Product
Frequently Bought Together
VWR® Stripping Buffers
Supplier: VWR
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
Amersham ECL Western Blotting Select Detection Reagent, Cytiva
Supplier: Cytiva
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Unstained Protein Standard, Broad Range, New England Biolabs
Supplier: New England Biolabs (NEB)
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Tris HCl pH 8.5, 1.0 M
Supplier: Rockland Immunochemical
DNA Wash Buffer, Concentrate, Zymo Research
Supplier: Zymo Research
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
BLOK™ Casein, 1% Casein Protein Blocking Solution, G-Biosciences
Supplier: G-Biosciences
Proteomelab™ SDS-MW Analysis Kit, SCIEX
Supplier: 0000013719
TEV PROTEASE HIS >95% 10000IU
Supplier: Genscript
PI MARKER ICE280/ICE3 7.05 200UL
Supplier: 0000014126
FACS and IF Staining Buffer with BSA
Supplier: Rockland Immunochemical
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript