ANTIBODY FLOTILLIN 1 A15114 100UL
Catalog # 77964-984
Supplier: ANTIBODIES.COM LLC
New Product
ANTIBODY FLOTILLIN 1 A15114 100UL
Catalog # 77964-984
Supplier: ANTIBODIES.COM LLC
Supplier Number: A15114-100
New Product
Frequently Bought Together
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Proteomelab™ SDS-MW Analysis Kit, SCIEX
Supplier: 0000013719
73254 METFORMIN HYDROCHLORIDE 5G
Supplier: STEMCELL Technologies
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
Supplier: Invitrogen
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
DNA Wash Buffer, Concentrate, Zymo Research
Supplier: Zymo Research
KIT PCR ASSAY ELISA HU PAPPA -20C 2PLATE
Supplier: 0000014373
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
SOLUTION ATTUNE WASH 250ML
Supplier: Invitrogen
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
Supplier: Invitrogen
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Chemi Blot Blocking Buffer, Azure
Supplier: Azure Biosystems
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
Supplier: Meso Scale Diagnostics
PI MARKER ICE280/ICE3 7.05 200UL
Supplier: 0000014126