ANTIBODY ANTI-FAF1 A14254 100UL
Catalog # 77964-320
Supplier: ANTIBODIES.COM LLC
New Product
ANTIBODY ANTI-FAF1 A14254 100UL
Catalog # 77964-320
Supplier: ANTIBODIES.COM LLC
Supplier Number: A14254-100
New Product
Frequently Bought Together
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
Supplier: Meso Scale Diagnostics
Rat Plasma, Equitech- Bio
Supplier: Equitech-Bio
PI MARKER 9.50 200UL
Supplier: 0000014126
DEFIBRINATED BLOOD SHEEP 100ML
Supplier: LAMPIRE BIOLOGICAL LAB MS
KIT PCR ASSAY ELISA HU PAPPA -20C 2PLATE
Supplier: 0000014373
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
73884 DIBUTYRYL CAMP SODIUM SALT 100MG
Supplier: STEMCELL Technologies
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
Supplier: Invitrogen
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
ASSAY IMMUNO CEFRAS GLOBAL CD8 HU 2VIAL
Supplier: MABTECH INC MS
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
Supplier: Invitrogen
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
LIGAND 646 HALOTAG 5UG LMAGING LIVE CELL
Supplier: Promega Corporation
100 0251 ELTROMBOPAG 5MG
Supplier: STEMCELL Technologies
ASSAY TAQMAN/ GENE EXPRESSION FAM 75RXN
Supplier: Invitrogen
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
BUFFER STABILIZR 7.5% DTPA-PURI BSA 50ML
Supplier: Revvity Health Sciences