ELISA KIT THROMBOSPONDIN 1 A2072 96 TEST
Catalog # 77963-306
Supplier: ANTIBODIES.COM LLC
New Product
ELISA KIT THROMBOSPONDIN 1 A2072 96 TEST
Catalog # 77963-306
Supplier: ANTIBODIES.COM LLC
Supplier Number: A2072-96
New Product
Frequently Bought Together
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
Tris HCl pH 8.5, 1.0 M
Supplier: Rockland Immunochemical
DNA Wash Buffer, Concentrate, Zymo Research
Supplier: Zymo Research
DEFIBRINATED BLOOD SHEEP 100ML
Supplier: LAMPIRE BIOLOGICAL LAB MS
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Proteomelab™ SDS-MW Analysis Kit, SCIEX
Supplier: 0000013719
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
LIGAND 646 HALOTAG 5UG LMAGING LIVE CELL
Supplier: Promega Corporation
Bovine Plasma with Citrate, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
FlashGel™ DNA, RNA and Recovery Systems
Supplier: Lonza
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
PI MARKER ICE280/ICE3 7.05 200UL
Supplier: 0000014126
Rat Plasma, Equitech- Bio
Supplier: Equitech-Bio
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript