ANTIBODY ANTI-GABRA4 A17286 100UL
Catalog # 77962-669
Supplier: ANTIBODIES.COM LLC
New Product
ANTIBODY ANTI-GABRA4 A17286 100UL
Catalog # 77962-669
Supplier: ANTIBODIES.COM LLC
Supplier Number: A17286-100
New Product
Frequently Bought Together
KIT PCR ASSAY ELISA HU PAPPA -20C 2PLATE
Supplier: 0000014373
73884 DIBUTYRYL CAMP SODIUM SALT 100MG
Supplier: STEMCELL Technologies
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
Supplier: Invitrogen
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
KIT BD-TAU ADVANTAGE PLUS
Supplier: QUANTERIX CORPORATION MS
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT HUMAN GLP-1R REPORTER 3X32ASSAYS
Supplier: INDIGO BIOSCIENCES INC MS
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
ASSAY IMMUNO CEFRAS GLOBAL CD8 HU 2VIAL
Supplier: MABTECH INC MS
100 0249 FORSKOLIN 25MG
Supplier: STEMCELL Technologies
Goat Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
Supplier: Invitrogen
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
Supplier: Meso Scale Diagnostics
Human Recombinant Shh (C24II)
Supplier: STEMCELL Technologies
100 0544 ERASTIN
Supplier: STEMCELL Technologies
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
100 0251 ELTROMBOPAG 5MG
Supplier: STEMCELL Technologies
BLOCKER AQB013 AQUAPORIN 5MG
Supplier: FOCUS BIOMOLECULES MS