ELISA KIT TF RECEPTOR A4497 48 TEST
Catalog # 77961-319
Supplier: ANTIBODIES.COM LLC
New Product
ELISA KIT TF RECEPTOR A4497 48 TEST
Catalog # 77961-319
Supplier: ANTIBODIES.COM LLC
Supplier Number: A4497-48
New Product
Frequently Bought Together
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
PI MARKER ICE280/ICE3 7.05 200UL
Supplier: 0000014126
Rat Plasma, Equitech- Bio
Supplier: Equitech-Bio
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Proteomelab™ SDS-MW Analysis Kit, SCIEX
Supplier: 0000013719
PI MARKER 9.50 200UL
Supplier: 0000014126
DEFIBRINATED BLOOD SHEEP 100ML
Supplier: LAMPIRE BIOLOGICAL LAB MS
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
MARKER 9.22 ICE280 ICE3 PI
Supplier: 0000014126
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
MOUSE SERUM US ORIGIN FOR COGLTN STUDIES
Supplier: Innovative Research Inc
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex