ANTIBODY ANTI-PAK5 A91289 100UL
Catalog # 77960-541
Supplier: ANTIBODIES.COM LLC
New Product
ANTIBODY ANTI-PAK5 A91289 100UL
Catalog # 77960-541
Supplier: ANTIBODIES.COM LLC
Supplier Number: A91289-100
New Product
Frequently Bought Together
DEFIBRINATED BLOOD SHEEP 100ML
Supplier: LAMPIRE BIOLOGICAL LAB MS
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
Supplier: Invitrogen
ASSAY IMMUNO CEFRAS GLOBAL CD8 HU 2VIAL
Supplier: MABTECH INC MS
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
BLOCKER AQB013 AQUAPORIN 5MG
Supplier: FOCUS BIOMOLECULES MS
KIT HUMAN GIPR REPORTER 3X32ASSAYS 96WL
Supplier: INDIGO BIOSCIENCES INC MS
Bovine Plasma with Citrate, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
74042 PHORBOL12 MYRISTATE 13 ACETATE 1MG
Supplier: STEMCELL Technologies
100 1042 CHIR99021
Supplier: STEMCELL Technologies
KIT ELISA ALT COMPLEMNT PATHWY 100UL/WEL
Supplier: Hycult Biotech
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
Supplier: Invitrogen
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
Supplier: Meso Scale Diagnostics
Human Recombinant Shh (C24II)
Supplier: STEMCELL Technologies
100 0544 ERASTIN
Supplier: STEMCELL Technologies