ANTIBODY ANTI- AF-1 A91581 100UL
Catalog # 77960-755
Supplier: ANTIBODIES.COM LLC
New Product
ANTIBODY ANTI- AF-1 A91581 100UL
Catalog # 77960-755
Supplier: ANTIBODIES.COM LLC
Supplier Number: A91581-100
New Product
Frequently Bought Together
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
73884 DIBUTYRYL CAMP SODIUM SALT 100MG
Supplier: STEMCELL Technologies
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
MOUSE SERUM US ORIGIN FOR COGLTN STUDIES
Supplier: Innovative Research Inc
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
BLOCKER AQB013 AQUAPORIN 5MG
Supplier: FOCUS BIOMOLECULES MS
HTRF STREPTAVIDIN TB-CONJUGATE 1000ASSAY
Supplier: Revvity Health Sciences
Bovine Plasma with Citrate, Equitech- Bio, Inc.
Supplier: Equitech-Bio
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
Supplier: Meso Scale Diagnostics
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT HUMAN GLP-1R REPORTER 3X32ASSAYS
Supplier: INDIGO BIOSCIENCES INC MS
BUFFER STABILIZR 7.5% DTPA-PURI BSA 50ML
Supplier: Revvity Health Sciences
73254 METFORMIN HYDROCHLORIDE 5G
Supplier: STEMCELL Technologies
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
Supplier: Invitrogen
100 0249 FORSKOLIN 25MG
Supplier: STEMCELL Technologies
ASSAY IMMUNO CEFRAS GLOBAL CD8 HU 2VIAL
Supplier: MABTECH INC MS
SOLUTION ATTUNE WASH 250ML
Supplier: Invitrogen
KIT ELISA ALT COMPLEMNT PATHWY 100UL/WEL
Supplier: Hycult Biotech
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
Supplier: Invitrogen