ANTIBODY ANTI-RHOU A91751 100UL
Catalog # 77960-851
Supplier: ANTIBODIES.COM LLC
New Product
ANTIBODY ANTI-RHOU A91751 100UL
Catalog # 77960-851
Supplier: ANTIBODIES.COM LLC
Supplier Number: A91751-100
New Product
Frequently Bought Together
Pharmalytes® IEF Carrier Ampholytes, Cytiva
Supplier: Cytiva
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
Supplier: Meso Scale Diagnostics
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Proteomelab™ SDS-MW Analysis Kit, SCIEX
Supplier: 0000013719
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
Tris HCl pH 8.5, 1.0 M
Supplier: Rockland Immunochemical
DNA Wash Buffer, Concentrate, Zymo Research
Supplier: Zymo Research
PI MARKER 5.85 200 UL
Supplier: 0000014126
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
SOLUTION ATTUNE WASH 250ML
Supplier: Invitrogen
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
Supplier: Invitrogen
Unstained Protein Standard, Broad Range, New England Biolabs
Supplier: New England Biolabs (NEB)
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
Supplier: Invitrogen