ANTIBODY ANTI- ACP1 A88590 100UL
Catalog # 77958-321
Supplier: ANTIBODIES.COM LLC
New Product
ANTIBODY ANTI- ACP1 A88590 100UL
Catalog # 77958-321
Supplier: ANTIBODIES.COM LLC
Supplier Number: A88590-100
New Product
Frequently Bought Together
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Bovine Plasma with Citrate, Equitech- Bio, Inc.
Supplier: Equitech-Bio
100 1042 CHIR99021
Supplier: STEMCELL Technologies
KIT BD-TAU ADVANTAGE PLUS
Supplier: QUANTERIX CORPORATION MS
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
MOUSE SERUM US ORIGIN FOR COGLTN STUDIES
Supplier: Innovative Research Inc
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT HUMAN GLP-1R REPORTER 3X32ASSAYS
Supplier: INDIGO BIOSCIENCES INC MS
ASSAY TAQMAN/ GENE EXPRESSION FAM 75RXN
Supplier: Invitrogen
KIT HUMAN GIPR REPORTER 3X32ASSAYS 96WL
Supplier: INDIGO BIOSCIENCES INC MS
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
Supplier: Invitrogen
100 0249 FORSKOLIN 25MG
Supplier: STEMCELL Technologies
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
ASSAY IMMUNO CEFRAS GLOBAL CD8 HU 2VIAL
Supplier: MABTECH INC MS
SOLUTION ATTUNE WASH 250ML
Supplier: Invitrogen
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
Supplier: Invitrogen
KIT ELISA ALT COMPLEMNT PATHWY 100UL/WEL
Supplier: Hycult Biotech
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
Supplier: Meso Scale Diagnostics