ELISA KIT RAT GRANULIN A6806 96 TEST
Catalog # 77955-984
Supplier: ANTIBODIES.COM LLC
New Product
ELISA KIT RAT GRANULIN A6806 96 TEST
Catalog # 77955-984
Supplier: ANTIBODIES.COM LLC
Supplier Number: A6806-96
New Product
Frequently Bought Together
VWR® Stripping Buffers
Supplier: VWR
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
LIGAND 646 HALOTAG 5UG LMAGING LIVE CELL
Supplier: Promega Corporation
Goat Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
Tris HCl pH 8.5, 1.0 M
Supplier: Rockland Immunochemical
DNA Wash Buffer, Concentrate, Zymo Research
Supplier: Zymo Research
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
Rat Plasma, Equitech- Bio
Supplier: Equitech-Bio