ELISA KIT MOUSE DNMT3B A5156 96 TEST
77949-743
New Product
Frequently Bought Together
Rat Plasma, Equitech- Bio
: Equitech-Bio
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
: Genetex
DEFIBRINATED BLOOD SHEEP 100ML
: LAMPIRE BIOLOGICAL LAB MS
Chicken Embryo Extract, MP Biomedical
: MP Biomedicals
Fixation, (buffer), Enzo Life Sciences
: Enzo Life Sciences
MOUSE SERUM US ORIGIN FOR COGLTN STUDIES
: Innovative Research Inc
Canine Plasma, Equitech- Bio, Inc.
: Equitech-Bio
Bovine Plasma with Citrate, Equitech- Bio, Inc.
: Equitech-Bio
Nano-Glo® Luciferase Assay System, Promega, Promega
: Promega Corporation
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
: New England Biolabs (NEB)
BUFFER JAW SSC 2X 1L PACKS
: G-Biosciences
LIGAND 646 HALOTAG 5UG LMAGING LIVE CELL
: Promega Corporation
Fixation/Permeabilization, (buffer), Enzo Life Sciences
: Enzo Life Sciences
Bis-Tris Mini Protein Gels, GenScript®
: Genscript