ELISA KIT MOUSE SYNTAXIN 1A A5804 48TEST
Catalog # 77950-017
Supplier: ANTIBODIES.COM LLC
New Product
ELISA KIT MOUSE SYNTAXIN 1A A5804 48TEST
Catalog # 77950-017
Supplier: ANTIBODIES.COM LLC
Supplier Number: A5804-48
New Product
Frequently Bought Together
DEFIBRINATED BLOOD SHEEP 100ML
Supplier: LAMPIRE BIOLOGICAL LAB MS
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
Blot Boxes, IBI Scientific
Supplier: IBI Scientific
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
LIGAND 646 HALOTAG 5UG LMAGING LIVE CELL
Supplier: Promega Corporation
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Bovine Plasma with Citrate, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
g-DNA Wash Buffer, Zymo Research
Supplier: Zymo Research
Rat Plasma, Equitech- Bio
Supplier: Equitech-Bio
AGAROSE ALFA SELECTOR ST 2ML
Supplier: NANOTAG BIOTECHNOLOGIES GMBH MS
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
Goat Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio