BROMPERIDOL DECANOATE HCL-25MG
Catalog # 77949-005
Supplier: TLC Standards
New Product
BROMPERIDOL DECANOATE HCL-25MG
Catalog # 77949-005
Supplier: TLC Standards
Supplier Number: B-071004-25MG
New Product
Frequently Bought Together
Sodium chloride 5 M, sterile-filtered
Supplier: Teknova
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
VWR® Acrylamide and Bis-Acrylamide
Supplier: VWR
PI MARKER ICE280/ICE3 4.65 200UL
Supplier: 0000014126
CUBIC-L Tissue-Clearing Reagent, for Animals
Supplier: TCI America
Carbon disulfide 99.9% for spectroscopy
Supplier: Thermo Scientific Chemicals
Acetonitrile 99.9%, meets analytical specification of Ph.Eur. for HPLC, far-UV
Supplier: Thermo Scientific Chemicals
Dimethyl sulfoxide 99.8+% for HPLC
Supplier: Thermo Scientific Chemicals
TISSUE-CLEARING RGNT CUBIC-B T3780-100ML
Supplier: TCI America
MARKER 9.22 ICE280 ICE3 PI
Supplier: 0000014126
TISSUECLEARING RGT CUBIC-R+M T3741-100ML
Supplier: TCI America
n-Pentane, anhydrous 99+%, Extra Pure
Supplier: Thermo Scientific Chemicals
N-Methyl-2-pyrrolidone (NMP) 99+% ACS
Supplier: Thermo Scientific Chemicals
Methanol 99.9% for analysis
Supplier: Thermo Scientific Chemicals
Dimethyl sulfoxide 99.7%, pure
Supplier: Thermo Scientific Chemicals
Lithium chloride 99+% ACS
Supplier: Thermo Scientific Chemicals