ELISA MOUSE NEPHRONECTIN A5282 96TEST
Catalog # 77949-995
Supplier: ANTIBODIES.COM LLC
New Product
ELISA MOUSE NEPHRONECTIN A5282 96TEST
Catalog # 77949-995
Supplier: ANTIBODIES.COM LLC
Supplier Number: A5282-96
New Product
Frequently Bought Together
Amersham ECL Western Blotting Select Detection Reagent, Cytiva
Supplier: Cytiva
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Proteomelab™ SDS-MW Analysis Kit, SCIEX
Supplier: 0000013719
PI MARKER 9.50 200UL
Supplier: 0000014126
PI MARKER MAURICE CIEF 4.05 210UL
Supplier: 0000014126
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
LIGAND 646 HALOTAG 5UG LMAGING LIVE CELL
Supplier: Promega Corporation
MARKER 6.14 MAURICE CIEF PI 210UL
Supplier: 0000014126
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Radiance Q Chemiluminescent Substrate, Azure Biosystems
Supplier: Azure Biosystems
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
Rat Plasma, Equitech- Bio
Supplier: Equitech-Bio
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript