ELISA KIT MOUSE BMPER A5741 96 TEST
Catalog # 77947-455
Supplier: ANTIBODIES.COM LLC
New Product
ELISA KIT MOUSE BMPER A5741 96 TEST
Catalog # 77947-455
Supplier: ANTIBODIES.COM LLC
Supplier Number: A5741-96
New Product
Frequently Bought Together
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Bovine Plasma with Citrate, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
DEFIBRINATED BLOOD SHEEP 100ML
Supplier: LAMPIRE BIOLOGICAL LAB MS
Chicken Embryo Extract, MP Biomedical
Supplier: MP Biomedicals
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
LIGAND 646 HALOTAG 5UG LMAGING LIVE CELL
Supplier: Promega Corporation
Unstained Protein Standard, Broad Range, New England Biolabs
Supplier: New England Biolabs (NEB)
MOUSE SERUM US ORIGIN FOR COGLTN STUDIES
Supplier: Innovative Research Inc
Fixation, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
Rat Plasma, Equitech- Bio
Supplier: Equitech-Bio
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
Goat Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio