ANTIBODY VGF RECOMBINAN RABBIT 100UL
Catalog # MSPP-ORB74992
Supplier: Biorbyt
ANTIBODY VGF RECOMBINAN RABBIT 100UL
Catalog # MSPP-ORB74992
Supplier: Biorbyt
Supplier Number: ORB74992
Frequently Bought Together
FLAG EPITOPE TAGDYKDDDDKAB HRP CONJUGA
Supplier: Invitrogen
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
Supplier: Genetex
ASSAY IMMUNO CEFRAS GLOBAL CD8 HU 2VIAL
Supplier: MABTECH INC MS
BUFFER JAW SSC 2X 1L PACKS
Supplier: G-Biosciences
BLOCKER AQB013 AQUAPORIN 5MG
Supplier: FOCUS BIOMOLECULES MS
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Bovine Plasma with Citrate, Equitech- Bio, Inc.
Supplier: Equitech-Bio
KIT BD-TAU ADVANTAGE PLUS
Supplier: QUANTERIX CORPORATION MS
100 1042 CHIR99021
Supplier: STEMCELL Technologies
DEFIBRINATED BLOOD SHEEP 100ML
Supplier: LAMPIRE BIOLOGICAL LAB MS
KIT ELISA ALT COMPLEMNT PATHWY 100UL/WEL
Supplier: Hycult Biotech
SOLUTION ATTUNE WASH 250ML
Supplier: Invitrogen
ANTIBODY CD8 ALPHA FITC CONJUGATE 516759
Supplier: Invitrogen
MOUSE SERUM US ORIGIN FOR COGLTN STUDIES
Supplier: Innovative Research Inc
ANTIBODY MSD GOLD SULFOTAG ANTI HUM 80UG
Supplier: Meso Scale Diagnostics
Fixation/Permeabilization, (buffer), Enzo Life Sciences
Supplier: Enzo Life Sciences
ASSAY TAQMAN/ GENE EXPRESSION FAM 75RXN
Supplier: Invitrogen
KIT HUMAN GLP-1R REPORTER 3X32ASSAYS
Supplier: INDIGO BIOSCIENCES INC MS
72262 ALL TRANS RETINOIC ACID 50MG
Supplier: STEMCELL Technologies