ELISA KIT PORCINE LBP A7472 48 TEST
77945-729
New Product
Frequently Bought Together
VWR® Stripping Buffers
: VWR
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
: New England Biolabs (NEB)
Nano-Glo® Luciferase Assay System, Promega, Promega
: Promega Corporation
Unstained Protein Standard, Broad Range, New England Biolabs
: New England Biolabs (NEB)
Fixation, (buffer), Enzo Life Sciences
: Enzo Life Sciences
DNA Wash Buffer, Concentrate, Zymo Research
: Zymo Research
Chicken Embryo Extract, MP Biomedical
: MP Biomedicals
BUFFER JAW SSC 2X 1L PACKS
: G-Biosciences
LIGAND 646 HALOTAG 5UG LMAGING LIVE CELL
: Promega Corporation
PI MARKER ICE280/ICE3 7.05 200UL
: 0000014126
SUBSTRATE TRIDENT PICO WESTERN HRP 500ML
: Genetex
Fixation/Permeabilization, (buffer), Enzo Life Sciences
: Enzo Life Sciences
Proteomelab™ SDS-MW Analysis Kit, SCIEX
: 0000013719
Bis-Tris Mini Protein Gels, GenScript®
: Genscript