TISSUE ARRAY MALIGNANT MELANOM 24CS/24CR
Catalog # MSPP-ME242C
Supplier: TISSUEARRAY.COM LLC MS
TISSUE ARRAY MALIGNANT MELANOM 24CS/24CR
Catalog # MSPP-ME242C
Supplier: TISSUEARRAY.COM LLC MS
Supplier Number: ME242C
Frequently Bought Together
Immobiline™ DryStrip Gels, Cytiva
Supplier: Cytiva
VWR® VividPro™ Fluorescent Stain
Supplier: VWR
Buffer P2 (Green), Zymo Research
Supplier: Zymo Research
DNA Elution Buffer, Zymo Research
Supplier: Zymo Research
Accessories for eBlot® L1 Fast Wet Transfer System for Mini Gels
Supplier: Genscript
E. coli GenLysate™ Bacterial Lysate, G-Biosciences
Supplier: G-Biosciences
Blot Boxes, IBI Scientific
Supplier: IBI Scientific
Pierce™ Protein A/G Magnetic Agarose, Thermo Scientific
Supplier: Invitrogen
Mammalian GenLysate™ Mouse Total Protein Lysates, G-Biosciences
Supplier: G-Biosciences
eStain™ L1 Protein Staining Device, GenScript
Supplier: Genscript
Mix & Go! 2X Stock Wash Buffer, Zymo Research
Supplier: Zymo Research
Protein Molecular Weight Marker Mix, Rockland Immunchemicals
Supplier: Rockland Immunochemical
Mix & Go! Dilution Buffer, Zymo Research
Supplier: Zymo Research
RNA RECOVERY BUFFER 10 ML
Supplier: Zymo Research
Antibody Fab Fragmentation, G-Biosciences
Supplier: G-Biosciences
TBS pH 7.5, 10X, Rockland Immunochemicals
Supplier: Rockland Immunochemical
1X TBS pH 7.8
Supplier: Rockland Immunochemical
1X PBS Buffer with 1% Casein
Supplier: Rockland Immunochemical
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript