KIT ELISA IFN GAMMA QUANTIKINE QUICKIT
Catalog # MSPP-QK285
Supplier: R&D Systems
KIT ELISA IFN GAMMA QUANTIKINE QUICKIT
Catalog # MSPP-QK285
Supplier: R&D Systems
Supplier Number: QK285
Frequently Bought Together
Dimethyl sulfoxide ≥99.9%, Ultra Pure Grade
Supplier: VWR
VWR® Stripping Buffers
Supplier: VWR
Vi-CELL BLU Reagent Kits, Beckman Coulter
Supplier: Beckman Coulter
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
Disposable PD-10 Desalting Columns, Cytiva
Supplier: Cytiva
PI MARKER 9.50 200UL
Supplier: 0000014126
DNA Wash Buffer, Concentrate, Zymo Research
Supplier: Zymo Research
PI MARKER 5.85 200 UL
Supplier: 0000014126
BUFFER TBS BLOCKING READY-TO-USE 500ML
Supplier: Li-Cor
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
TAE 50X Buffer, MP Biomedicals
Supplier: MP Biomedicals
Radiance Q Chemiluminescent Substrate, Azure Biosystems
Supplier: Azure Biosystems
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
Proteomelab™ SDS-MW Analysis Kit, SCIEX
Supplier: 0000013719