BUFFER 10X TE140 PH 7.4
Catalog # MSPP-BUTE14010
Supplier: BRUKER SCIENTIFIC, LLC MS
BUFFER 10X TE140 PH 7.4
Catalog # MSPP-BUTE14010
Supplier: BRUKER SCIENTIFIC, LLC MS
Supplier Number: BU-TE-140-10
Frequently Bought Together
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
VWR® Disposable Transfer Pipettes
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
Dimethyl sulfoxide ≥99.9%, Ultra Pure Grade
Supplier: VWR
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Water, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Nano-Glo® Luciferase Assay System, Promega, Promega
Supplier: Promega Corporation
SYBR® Safe DNA Stain, 10,000 X, Edvotek
Supplier: Edvotek