a-Cyclodextrin USP
Product Details & Documents
a-Cyclodextrin is a cyclic oligosaccharide composed of seven glucose units, known for its unique ability to form inclusion complexes with various guest molecules. This property makes it an invaluable compound in the pharmaceutical, food, and cosmetic industries. Its ability to enhance the solubility and stability of poorly soluble drugs allows for improved bioavailability, making it a popular choice in drug formulation. Additionally, a-Cyclodextrin is utilized in food applications to encapsulate flavors and aromas, thereby enhancing product quality and shelf life. In cosmetics, it serves as a stabilizer and carrier for active ingredients, ensuring effective delivery and prolonged efficacy.
Moreover, a-Cyclodextrin's non-toxic and biodegradable nature aligns with the growing demand for sustainable and safe ingredients in various applications. Its versatility extends to research, where it is employed in analytical chemistry for separation and purification processes. With its broad range of applications and benefits, a-Cyclodextrin stands out as a crucial ingredient for professionals seeking to innovate and improve product formulations across multiple industries.
Store at RT.
- CAS Number:10016-20-3
- Formula:C₃₆H₆₀O₃₀
- MDL Number:MFCD00078207
- Molecular Weight:972.9 g/mol
- Storage Temperature:Room Temperature
Specifications
Specifications
- Pack type:Plastic bottle
- Size:5 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



