TISSUE ARRAY HU LUNG CARCINOMA NAT CS55
Catalog # MSPP-ODCTRSLUG
Supplier: TISSUEARRAY.COM LLC MS
TISSUE ARRAY HU LUNG CARCINOMA NAT CS55
Catalog # MSPP-ODCTRSLUG
Supplier: TISSUEARRAY.COM LLC MS
Supplier Number: OD-CT-RSLUG04-003
Frequently Bought Together
Mammalian GenLysate™ Human Total Protein Lysates, G-Biosciences
Supplier: G-Biosciences
HRP-PROTEIN L 1MG
Supplier: Genscript
E. coli GenLysate™ Bacterial Lysate, G-Biosciences
Supplier: G-Biosciences
DNA Elution Buffer, Zymo Research
Supplier: Zymo Research
Buffer P2 (Green), Zymo Research
Supplier: Zymo Research
Blot Boxes, IBI Scientific
Supplier: IBI Scientific
2-D Quant Kit, Cytiva
Supplier: Cytiva
IPG Buffers, Cytiva
Supplier: Cytiva
OSM (227AA) HUMAN >95% 50UG
Supplier: Genscript
Buffer P3 (Yellow), Zymo Research
Supplier: Zymo Research
GelBond® Film
Supplier: Lonza
1X PBS Buffer with 1% Casein
Supplier: Rockland Immunochemical
g-DNA Wash Buffer, Zymo Research
Supplier: Zymo Research
Antibody Fab Fragmentation, G-Biosciences
Supplier: G-Biosciences
2D Electrophoresis Reagents, G-Biosciences
Supplier: G-Biosciences
Pierce™ Ni-NTA Magnetic Agarose, Thermo Scientific
Supplier: Invitrogen
RNA RECOVERY BUFFER 10 ML
Supplier: Zymo Research
Blue Prestained Protein Standard, Broad Range (11 to 250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
KIT DENSITY MARKER BEADS 6 VIALS POLYETH
Supplier: COSPHERIC MS