ASSAY IMMUNO CEFRAS GLOBAL CD8 HU 2VIAL
Catalog # MSPP-3638-1
Supplier: MABTECH INC MS
ASSAY IMMUNO CEFRAS GLOBAL CD8 HU 2VIAL
Catalog # MSPP-3638-1
Supplier: MABTECH INC MS
Supplier Number: 3638-1
Frequently Bought Together
Ward's® Protein Fixative Stain
Supplier: Ward's Science
Sodium dodecyl sulfate (SDS) 10% in aqueous solution
Supplier: Rockland Immunochemical
OLIGO BINDING BUFFER (40 ML)
Supplier: Zymo Research
100-Place Microscope Slide Box
Supplier: Alkali Scientific
Western Blot Boxes
Supplier: Alkali Scientific
Electrophoresis Running Buffers, G-Biosciences
Supplier: G-Biosciences
Perfect Protein™ Markers, MilliporeSigma
Supplier: MilliporeSigma
Accessories for eBlot® L1 Fast Wet Transfer System for Mini Gels
Supplier: Genscript
FLUOTAG X2 ANTI ALFA ALEXAFLUOR 647 2ML
Supplier: NANOTAG BIOTECHNOLOGIES GMBH MS
Endo-Wash Buffer, Zymo Research
Supplier: Zymo Research
DNA BINDING BUFFER 100 ML
Supplier: Zymo Research
GelBond® Film
Supplier: Lonza
INHIBITOR YEAST 100X PROTEASEARREST 5ML
Supplier: G-Biosciences
CASTING TRAY FOR ELECTROPHORESIS SYSTEM
Supplier: G-Biosciences
RNA Prep Buffer, Zymo Research
Supplier: Zymo Research
Buffer P3 (Yellow), Zymo Research
Supplier: Zymo Research
EGTA 1M PH 8.0 STERILE 100ML
Supplier: 0000014548
PINPOINT BINDING BUFFER 6 ML F/ISOLATION
Supplier: Zymo Research
ScanLater™ Western Blot Assay Kit, Molecular Devices
Supplier: Molecular Devices
AGAROSE ALFA SELECTOR ST 2ML
Supplier: NANOTAG BIOTECHNOLOGIES GMBH MS