(2-Morpholinophenyl)methanol ≥95%
Glass bottle, 10 g
Catalog # 77903-399
New Product
Supplier: Ambeed
: A175119-10G
(2-Morpholinophenyl)methanol ≥95%
Catalog # 77903-399
Supplier: Ambeed
New Product
CAS Number: 465514-33-4
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
: for Research Use Only
Synonyms: (2-Morpholin-4-yl-phenyl)methanol
- CAS Number:465514-33-4
- Formula:C₁₁H₁₅NO₂
- MDL Number:MFCD03086181
- Molecular Weight:193.240 g/mol
Specifications
Specifications
- Pack type:Glass bottle
- Size:10 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



