- Ø:¹/₂"
- Description:Model IB, unit dose powder sampler, single sample design, 6 ports at 1.5", 7.5", 13.5", 19.5", 25.5", 31.5" from tip
- Length:3' (91.4 cm)
- Material:316 stainless steel
- Environmentally Preferable:
Unit-dose and bulk powder, liquid, and semi-solid samplers available in various lengths and configurations.
Precision sampling: Designed for accurate and repeatable sampling of powders from bulk containers, ensuring representative and consistent samples for quality control and testing
- Pharmaceutical-grade construction: Made from high-quality 316L stainless steel or FDA-compliant materials, ensuring compatibility with GMP environments and resistance to corrosion or contamination
- Ease of cleaning and sterilization: Engineered for quick disassembly and thorough cleaning, supporting clean-in-place (CIP) and autoclave sterilization procedures
- Customizable and scalable: Can be tailored to specific sampling applications with adjustable volumes and configurations, ideal for R and D, pilot, and production-scale environments
- Compliance ready: Supports regulatory compliance with FDA, USP, and ISO standards, making them suitable for validated sampling protocols in pharmaceutical and cleanroom operations
Global Pharma specializes in high-quality sampling solutions for the pharmaceutical, biotech, and related industries. The product portfolio includes unit dose and bulk powder, liquid, and semi-solid samplers, designed to meet stringent regulatory requirements. These samplers are available in a variety of configurations to ensure precise and contamination-free sample collection.
With a focus on innovation and compliance, Global Pharma’s samplers cater to diverse manufacturing environments, providing single-use and reusable options for enhanced process efficiency and sterility assurance. Their products are widely used in quality control, R and D, and production settings, making them a trusted choice for pharmaceutical manufacturers worldwide.
All samplers are supplied with a PVC die retrieval tool.