PROBE TEMPERATUR F/ICTD ANALOG I/O MODUL
Catalog # MSPP-94ICTD
Supplier: MOUSER ELECTRONICS MS
PROBE TEMPERATUR F/ICTD ANALOG I/O MODUL
Catalog # MSPP-94ICTD
Supplier: MOUSER ELECTRONICS MS
Supplier Number: 94-ICTD
Frequently Bought Together
High Speed Multi-Plate Shaker Vortex Mixer, MPS-1, Grant Instruments
Supplier: GRANT USA INC.
Cartridges for Water Purification Systems
Supplier: ELGA LabWater
VWR® Ultrasonic Cleaners
Supplier: VWR International
CERAMIC SPACER SET 611
Supplier: Corning
Accessories for Corning® LSE™ 49L Shaking Incubator, Corning
Supplier: Corning
CONNECTOR T PC SWIVEL MALE LUER 2 FEMALE
Supplier: 0000019222
Accessories for VWR® Vortex Mixers
Supplier: VWR International
VWR® ULTREX™ Acids Bottle Top Dispenser
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Laboratory Line Vertical Autoclaves, Heidolph Tuttnauer
Supplier: Heidolph NA, LLC
VWR® Standard Microplate Vortex Mixers
Supplier: VWR International
Nalgene Magnetic Carboy Stirrer, Thermo Scientific
Supplier: Thermo Fisher Scientific
Accessories for Duo™ Dual System, Ultrapure (18.2 MΩ-cm) and DI Lab Water, Avidity Sciences
Supplier: AVIDITY SCIENCE
Accessories for Corning® LSE™ Shaking Incubator, 71 L, Corning
Supplier: Corning
STIRRER MULTIPOINT TELEMODUL 40 C 1200ML
Supplier: Thermo Fisher Scientific
STAPLER CLASSIC DESKTOP BLK 30SHEET CAP
Supplier: STAPLES BUSINESS ADVANTAGE MS
VALVE FEMALE LUER LOCK INLET 0.433PSI
Supplier: 0000019222
VWR® Premium Laboratory Freezers
Supplier: VWR International
Q700 Sonicator without a Probe, Qsonica
Supplier: QSonica, LLC.