Sodium taurocholate hydrate ≥97%
Glass bottle, 5 g
Catalog # 77891-116
New Product
Supplier: Ambeed
Supplier #: A555499-5G
Sodium taurocholate hydrate ≥97%
Catalog # 77891-116
Supplier: Ambeed
New Product
CAS Number: 345909-26-4
Jump to:
Product Details & Documents
Caution: for Research Use Only
Synonyms: Taurocholic acid sodium salt hydrate, Cholaic acid sodium salt hydrate, Sodium 2-((R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethane-1-sulfonate hydrate, Sodium 2-((R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethane-1-sulphonate hydrate
- CAS Number:345909-26-4
- Formula:C₂₆H₄₆NNaO₈S
- MDL Number:MFCD00150819
- Molecular Weight:555.70 g/mol
Specifications
Specifications
- Pack type:Glass bottle
- Size:5 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



