Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SOLVENT RESERVOIR FILTER 1/8 ID 10UM
Catalog # 77889-310
Supplier:
Optimize
New Product
Expand image
SOLVENT RESERVOIR FILTER 1/8 ID 10UM
Catalog # 77889-310
Supplier:
Optimize
Supplier Number:
10-04-05523
New Product
Frequently Bought Together
Acrodisc® Syringe Filters for Prefiltration, Cytiva
Supplier: Cytiva
Acrodisc® Syringe Filters, 25 mm
Supplier: Cytiva
Syringe filters, Acrodisc®, Supor® membrane, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Acro® 37 TF Vent Devices, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
VWR® Syringe Filters
Supplier: VWR International
Whatman™ GD/X Syringe Filters, Prefilter, Non-Sterile, Whatman products (Cytiva)
Supplier: Cytiva
Acro® 50 Vent Filters, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Acrodisc® WBC (White Blood Cell) Syringe Filter, Cytiva (Formerly Pall Lab)
Supplier: Cytiva