Orders LinkContactUsLinkComponent
CytoFLEX Mosaic Detection Modules
CytoFLEX Mosaic 88 spectral detection module N-V-B-Y-R-I (6 laser CytoFLEX LX NUV)
Catalog # 77886-970
Supplier:   Beckman Coulter
 :  D19584
CytoFLEX Mosaic Detection Modules
Catalog # 77886-970
Supplier:  Beckman Coulter
CytoFLEX Mosaic Detection Modules
CytoFLEX Mosaic Detection Modules
CytoFLEX Mosaic 88 spectral detection module N-V-B-Y-R-I (6 laser CytoFLEX LX NUV)
Price
Inquire For Price
Log in for availability
Each
Product Details & Documents
The CytoFLEX Mosaic Detection Module is the natural evolution of the CytoFLEX platform. A smart approach to spectral analysis, the detection module connects to the CytoFLEX LX/S flow cytometer, providing the flexibility to mix and match different detection methods in one instrument.
  • A flexible approach to spectral flow cytometry
With up to 88 detection channels, the CytoFLEX Mosaic Detection Module enhances the CytoFLEX S or LX flow cytometer, empowering users to tackle the most complex experiments. Together, they create the most advanced and powerful CytoFLEX flow cytometry solution to date.
Specifications
  • Description:
    CytoFLEX Mosaic 88 spectral detection module N-V-B-Y-R-I (6 laser CytoFLEX LX NUV)