GEL PYROTELL 5ML/VIAL PK25
Catalog # MSPP-G500325
Supplier: ASSOCIATES OF CAPE COD INC MS
GEL PYROTELL 5ML/VIAL PK25
Catalog # MSPP-G500325
Supplier: ASSOCIATES OF CAPE COD INC MS
Supplier Number: G500325
Frequently Bought Together
Immobiline™ DryStrip Gels, Cytiva
Supplier: Cytiva
E. coli GenLysate™ Bacterial Lysate, G-Biosciences
Supplier: G-Biosciences
Blot Boxes, IBI Scientific
Supplier: IBI Scientific
Buffer P2 (Green), Zymo Research
Supplier: Zymo Research
Accessories for eBlot® L1 Fast Wet Transfer System for Mini Gels
Supplier: Genscript
Mix & Go! Dilution Buffer, Zymo Research
Supplier: Zymo Research
RIPA Lysis Buffer, Rockland Immunochemicals
Supplier: Rockland Immunochemical
ELISA KIT ARG8 VASOPRESSINS
Supplier: ARBOR ASSAYS MS
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
Zyppy Elution Buffer, Zymo Research
Supplier: Zymo Research
1X PBS Buffer with 1% Casein
Supplier: Rockland Immunochemical
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
Ammonium acetate
Supplier: G-Biosciences
Mammalian GenLysate™ Human Total Protein Lysates, G-Biosciences
Supplier: G-Biosciences
100-Place Microscope Slide Box
Supplier: Alkali Scientific
EGTA 1M PH 7.4 STERILE
Supplier: BIOWORLD MS
AGAROSE ALFA SELECTOR PE 2ML
Supplier: NANOTAG BIOTECHNOLOGIES GMBH MS
Borate Saline Buffered (BSB), Rockland Immunochemicals
Supplier: Rockland Immunochemical